missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ASCT2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA534201
Code : NEW
Additional Details : Weight : 0.01000kg
Description
Percent identity with other species by BLAST analysis: Human (100%) Gibbon, Monkey, Rat (93%) Marmoset, Hamster, Elephant, Horse (87%) Mouse, Bat, Bovine (80%).
SLC1A5 (Solute carrier family 1 (neutral amino acid transporter), member 5) is a member of the Na(+)-dependent amino acid transporter superfamily. It has a broad substrate specificity, a preference for zwitterionic amino acids, and a sodium-dependence. It accepts as substrates all neutral amino acids, including glutamine, asparagine, and branched-chain and aromatic amino acids, and excludes methylated amino acids, anionic amino acids, and cationic amino acids. It acts as a cell surface receptor for feline endogenous virus RD114, baboon M7 endogenous virus and type D simian retroviruses.Specifications
ASCT2 | |
Polyclonal | |
Unconjugated | |
SLC1A5 | |
AAAT; ASC-like Na(+)-dependent neutral amino acid transporter ASCT2; Asct2; ATB(0); ATB0; ATBO; baboon M7 virus receptor; CAZ-associated structural protein; H4-ASCT2; H4-system ASC-like transporter; insulin-activated amino acid transporter; M7V1; M7VS1; neutral amino acid transporter B; neutral amino acid transporter B(0); R16; RD114 virus receptor; RD114/simian type D retrovirus receptor; RDR; RDRC; SLC1A5; Slc1a7; sodium-dependent neutral amino acid transporter ASCT2; sodium-dependent neutral amino acid transporter type 2; solute carrier family 1 (neutral amino acid transporter), member 5; solute carrier family 1 member 5; solute carrier family 1, member 7 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
6510 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
Q15758 | |
SLC1A5 | |
Synthetic 15 amino acid peptide from C-Terminus of human SLC1A5. | |
50 μg | |
Primary | |
Human | |
Antibody | |
IgG |