missing translation for 'onlineSavingsMsg'
Learn More

Ascorbyl Palmitate, TRC

Product Code. 17920715
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17920715 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17920715 Supplier Toronto Research Chemicals Supplier No. TRCA78702025G
 Restricted Item
Estimated Shipment: 28-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 137-66-6
Molecular Formula C22 H38 O7
Quantity 25 g
Synonym L-Ascorbic acid, 6-palmitate (8CI), Palmitic acid, 6-ester with ascorbic acid (6CI,7CI), 6-Hexadecanoyl-L-ascorbic acid, 6-Monopalmitoyl-L-ascorbate, 6-O-Palmitoyl-L-ascorbic acid, 6-O-Palmitoyl-L-ascorbic acid, 6-O-Palmitoylascorbic acid, 6-Palmitate-L-ascorbic acid, 6-Palmitoylascorbic acid, AP, Ascorbic acid 6-palmitate, Ascorbic acid palmitate, Ascorbic palmitate, Ascorboyl palmitate, Ascorbyl 6-palmitate, Ascorbyl monopalmitate, Ascorbyl palmitate, Ascorbylpalmitic acid, Cetyl ascorbate, E 304, Grindox 562, L-Ascorbyl 6-palmitate, L-Ascorbyl monopalmitate, L-Ascorbyl palmitate, NSC 402451, Ondascora, Palmitoyl L-ascorbic acid, Quicifal, VCP 10, Vcpal
InChI Formula InChI=1S/C22H38O7/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-18(24)28-16-17(23)21-19(25)20(26)22(27)29-21/h17,21,23,25-26H,2-16H2,1H3/t17-,21+/m0/s1
SMILES CCCCCCCCCCCCCCCC(=O)OC[C@H](O)[C@H]1OC(=O)C(=C1O)O
IUPAC Name [(2S)-2-[(2R)-3,4-dihydroxy-5-oxo-2H-furan-2-yl]-2-hydroxyethyl] hexadecanoate
Molecular Weight (g/mol) 414.53
Formula Weight 414.2618
Chemical Name or Material Ascorbyl Palmitate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.