missing translation for 'onlineSavingsMsg'
Learn More

Ascorbyl palmitate, MedChemExpress

Product Code. 16468546
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
200 mg
500 mg
Unit Size:
100mg
200mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16468546 100 mg 100mg
16422236 500 mg 500mg
16411456 200 mg 200mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16468546 Supplier Medchem Express Supplier No. HYB0987100mg
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Ascorbyl palmitate is an ester formed from ascorbic acid and palmitic acid creating an vitamin C, it is also used as an antioxidant food additive.

Ascorbyl palmitate is an ester formed from ascorbic acid and palmitic acid creating an vitamin C, it is also used as an antioxidant food additive.

Specifications

Chemical Name or Material Ascorbyl palmitate
CAS 137-66-6
Color White
Purity Grade Notes Research
Molecular Formula C22H38O7
Quantity 100 mg
Synonym L-Ascorbic acid 6-hexadecanoate 6-O-Palmitoyl-L-ascorbic acid
Solubility Information DMSO : 100 mg/mL (241.24 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES OC([C@H](O1)[C@H](COC(CCCCCCCCCCCCCCC)=O)O)=C(O)C1=O
Molecular Weight (g/mol) 414.53
Formula Weight 414.53
Percent Purity 99.61%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.