missing translation for 'onlineSavingsMsg'
Learn More

Ascorbic Acid, Fine Powder, 200 to 325 Mesh, U.S.P. - F.C.C., J.T. Baker™

Product Code. 15577874 Shop All J.T.Baker Products
Click to view available options
Quantity:
500 g
1 kg
Packaging:
Poly Bottle
Unit Size:
1kg
500g
This item is not returnable. View return policy

Product Code. 15577874

Brand: Avantor J.T.Baker 093707

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 50-81-7
Molecular Formula C6H8O6
Molecular Weight (g/mol) 176.12
MDL Number MFCD00064328
InChI Key CIWBSHSKHKDKBQ-DOMZIZNONA-N
Synonym l-ascorbic acid, ascorbic acid, vitamin c, l-ascorbate, ascorbate, ascorbicap, l +-ascorbic acid, cevitamic acid, ascoltin, hybrin
PubChem CID 54670067
ChEBI CHEBI:29073
IUPAC Name (2R)-2-[(1S)-1,2-dihydroxyethyl]-3,4-dihydroxy-2H-furan-5-one
SMILES OC[C@H](O)[C@H]1OC(=O)C(O)=C1O

Specifications

Mesh Size 200 to 325 Mesh
Packaging Poly Bottle
Quantity 500 g
Specific Gravity 1L = 1.65kg
Formula Weight 176.12
Grade FCC/USP
Physical Form Fine Powder
Chemical Name or Material Ascorbic Acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.