missing translation for 'onlineSavingsMsg'
Learn More

Ascorbic Acid, Fine Granular, 20 to 80 Mesh, U.S.P. - F.C.C., J.T. Baker™

Product Code. 15597874
Click to view available options
Quantity:
125 g
500 g
1 kg
Packaging:
Poly Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 50-81-7
Molecular Formula C6H8O6
Molecular Weight (g/mol) 176.12
MDL Number MFCD00064328
InChI Key CIWBSHSKHKDKBQ-DOMZIZNONA-N
Synonym l-ascorbic acid, ascorbic acid, vitamin c, l-ascorbate, ascorbate, ascorbicap, l +-ascorbic acid, cevitamic acid, ascoltin, hybrin
PubChem CID 54670067
ChEBI CHEBI:29073
IUPAC Name (2R)-2-[(1S)-1,2-dihydroxyethyl]-3,4-dihydroxy-2H-furan-5-one
SMILES OC[C@H](O)[C@H]1OC(=O)C(O)=C1O

Specifications

Mesh Size 20 to 80 Mesh
Packaging Poly Bottle
Quantity 125 g
Specific Gravity 1L = 1.65kg
Formula Weight 176.12
Grade FCC/USP
Physical Form Fine Granular
Chemical Name or Material Ascorbic Acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'