missing translation for 'onlineSavingsMsg'
Learn More

Ascorbic Acid, ACS Reagent Grade, Ricca Chemical

Product Code. 31750975
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
31750975 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 31750975 Supplier Ricca Chemical Company Supplier No. RDCA0750100B1
 Restricted Item
Estimated Shipment: 14-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Specifications

CAS 50-81-7
CAS Min % 98.0
CAS Max % 100.0
Color White
Packaging White Poly Wide Mouth Bottle
Molecular Formula C6H8O6
MDL Number MFCD00064328
Quantity 100 g
Synonym l-ascorbic acid, ascorbic acid, vitamin c, l-ascorbate, ascorbate, ascorbicap, l +-ascorbic acid, cevitamic acid, ascoltin, hybrin
InChI Key CIWBSHSKHKDKBQ-DOMZIZNONA-N
SMILES OC[C@H](O)[C@H]1OC(=O)C(O)=C1O
IUPAC Name (5R)-5-[(1S)-1,2-dihydroxyethyl]-3,4-dihydroxy-2,5-dihydrofuran-2-one
Molecular Weight (g/mol) 176.12
PubChem CID 54670067
ChEBI CHEBI:29073
Percent Purity Min. 99%
Grade ACS
Physical Form Crystals or Powder
Chemical Name or Material Ascorbic Acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.