missing translation for 'onlineSavingsMsg'
Learn More

Ascomycin, MedChemExpress

Product Code. 16495748
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16495748 10 mg 10mg
16491907 50 mg 50mg
16466226 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16495748 Supplier Medchem Express Supplier No. HY13557
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Ascomycin (Immunomycin; FR-900520; FK520) is an ethyl analog of Tacrolimus (FK506) with strong immunosuppressant properties. Ascomycin is also a macrocyclic polyketide antibiotic with multiple biological activities such as anti-malarial, anti-fungal and anti-spasmodic. Ascomycin prevents graft rejection and has potential for varying skin ailments research.

Ascomycin (Immunomycin

Specifications

Chemical Name or Material Ascomycin
Physical Form Powder
CAS 104987-12-4
Color White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C43H69NO12
For Use With (Application) COVID-19-immunoregulation
Synonym Immunomycin FR-900520 FK520
Solubility Information DMSO : ≥ 39 mg/mL (49.24 mM)
SMILES O=C([C@@](CCCC1)([H])N1C(C([C@@]2(O)[C@H](C)C[C@H](OC)[C@@](O2)([H])[C@@H](OC)C[C@@H](C)C/C(C)=C/[C@H]3CC)=O)=O)O[C@H](/C(C)=C/[C@H]4C[C@@H](OC)[C@H](O)CC4)[C@H](C)[C@@H](O)CC3=O
Molecular Weight (g/mol) 792.01
Formula Weight 792.01
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.