missing translation for 'onlineSavingsMsg'
Learn More

Ascomycin, 95%

Product Code. 15488229
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15488229 100 mg 100mg
15478229 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15488229 Supplier Thermo Scientific Alfa Aesar Supplier No. J66751.MC
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Internal standard for LC-MS/MS testing of the immunosuppressants tacrolimus, sirolimus, and everolimus. | CAS: 104987-12-4 | C43H69NO12 | 792.02 g/mol

Ascomycin is a potent immunosuppressive agent that could be used as a potential therapeutic agent for autoimmune diseases. Ascomycin is an inhibitor of PP2. It is an internal standard for LC-MS/MS testing of the immunosuppressants tacrolimus, sirolimus, and everolimus. This Certified Spiking Solution™ is suitable for critical quantitative applications in clinical and diagnostic testing such as therapeutic drug monitoring assays to ensure patients remain within the narrow therapeutic range of these immunosuppressants.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ascomycin is a potent immunosuppressive agent that could be used as a potential therapeutic agent for autoimmune diseases. Ascomycin is an inhibitor of PP2. It is an internal standard for LC-MS/MS testing of the immunosuppressants tacrolimus, sirolimus, and everolimus. This Certified Spiking Solution® is suitable for critical quantitative applications in clinical and diagnostic testing such as therapeutic drug monitoring assays to ensure patients remain within the narrow therapeutic range of these immunosuppressants.

Solubility
Soluble in DMSO at 65mg/ml; soluble in ethanol at 50mg/m; with warming. Very poorly soluble in water

Notes
Research Use only: Not intended for animal or human diagnostic or therapeutic use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104987-12-4
Molecular Formula C43H69NO12
Molecular Weight (g/mol) 792.02
MDL Number MFCD06198665
InChI Key ZDQSOHOQTUFQEM-NURRSENYSA-N
Synonym ascomycin, immunomycin, unii-auf4u5nsjk, ascomycin fk520, auf4u5nsjk, chembl8597, 15,19-epoxy-3h-pyrido 2,1-c 1,4 oxaazacyclotricosine-1,7,20,21 4h,23h-tetrone, 8-ethyl-5,6,8,11,12,13,14,15,16,17,18,19,24,25,26,26a-hexadecahydro-5,19-dihydroxy-3-2-4-hydroxy-3-methoxycyclohexyl-1-methylethenyl-14,16-dimethoxy-4,10,12,18-tetram, ascomycin from streptomyces hygroscopicus var. ascomyceticus, 15,19-epoxy-3h-pyrido 2,1-c 1,4 oxaazacyclotricosine-1,7,20,21 4h,23h-tetrone, 5,6,8,11,12,13,14,15,16,17,18,19,24,25,26,26a-hexadecahydro-5,19-dihydroxy-8-ethyl-3-2-4-hydroxy-3-methoxycyclohexyl-1-methylethenyl-14,16-dimethoxy-4,10,12,18-tetramethyl-, 3s-3r* e 1s*,3s*,4s* ,4s*,19s*,26ar*, ascomycin fk 520, fr 900520, imm
PubChem CID 5282071
ChEBI CHEBI:29582
SMILES CCC1C=C(CC(CC(C2C(CC(C(O2)(C(=O)C(=O)N3CCCCC3C(=O)OC(C(C(CC1=O)O)C)C(=CC4CCC(C(C4)OC)O)C)O)C)OC)OC)C)C

Specifications

Melting Point 156°C to 160°C
Boiling Point 868°C
Flash Point >110°C (230°F)
Packaging Plastic bottle
Quantity 100 mg
Solubility Information Soluble in DMSO at 65mg/ml; soluble in ethanol at 50mg/m; with warming. Very poorly soluble in water
Formula Weight 790
Percent Purity 95%
Physical Form Powder
Chemical Name or Material Ascomycin

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.