missing translation for 'onlineSavingsMsg'
Learn More

ASCL4 Antibody - Azide and BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody

£151.00 - £359.00

Specifications

Antigen ASCL4
Dilution Western Blot 1:500 - 1:1000, Immunohistochemistry 1:50-1:100, Immunohistochemistry-Paraffin
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18642992
View Documents
Novus Biologicals
NBP2-92741-0.02ml
0.02 mL
£151.00
0.02mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18613222
View Documents
Novus Biologicals
NBP2-92741-0.1ml
0.1 mL
£359.00
0.10mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

ASCL4 Polyclonal antibody specifically detects ASCL4 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications

Specifications

ASCL4
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Human, Mouse, Rat
achaete-scute complex homolog 4 (Drosophila), bHLHa44, class A basic helix-loop-helix protein 44, class II bHLH protein ASCL4
Recombinant fusion protein containing a sequence corresponding to amino acids 1-70 of human ASCL4 (NP_982260.3). METRKPAERLALPYSLRTAPLGVPGTLPGLPRRDPLRVALRLDAACWEWARSGCARGWQYLPVPLDSAFE
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Western Blot 1:500 - 1:1000, Immunohistochemistry 1:50-1:100, Immunohistochemistry-Paraffin
Polyclonal
Purified
RUO
PBS with 50% glycerol, pH7.3.
121549
IgG
Affinity purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.