missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ASCL3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA568843
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 92%; Dog: 85%; Guinea Pig: 85%; Horse: 77%; Human: 85%; Mouse: 100%; Rat: 93%.
ASCL3 is a protein coding gene, transcriptional repressor, and inhibits myogenesis.
Specifications
| ASCL3 | |
| Polyclonal | |
| Unconjugated | |
| ASCL3 | |
| achaete-scute complex homolog 3; achaete-scute complex homolog 3 (Drosophila); achaete-scute complex homolog-like 3; achaete-scute family bHLH transcription factor 3; achaete-scute homolog 3; Ascl3; ASH-3; ASH3A; bHLH transcription factor Sgn-1 (Salivary Glands 1); bHLH transcriptional regulator Sgn-1; bHLHa42; class A basic helix-loop-helix protein 42; HASH3; ICRFP703B1614Q5.4; ICRFP703N2430Q5.4; mASH3; mASH-3; Sgn1 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 56787 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9JJR7 | |
| ASCL3 | |
| synthetic peptide directed towards the middle region of mouse ASCL3. | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction