missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ASCL2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA541489
This item is not returnable.
View return policy
Description
Peptide sequence: RPQAVRPSAP RGPPGTTPVA ASPSRASSSP GRGGSSEPGS PRSAYSSDDS Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 93%; Human: 100%; Mouse: 79%; Pig: 100%; Rat: 93%.
This gene is a member of the basic helix-loop-helix (BHLH) family of transcription factors. It activates transcription by binding to the E box (5'-CANNTG-3'). Dimerization with other BHLH proteins is required for efficient DNA binding. Involved in the determination of the neuronal precursors in the peripheral nervous system and the central nervous system.
Specifications
| ASCL2 | |
| Polyclonal | |
| Unconjugated | |
| ASCL2 | |
| 2410083I15Rik; achaete scute-like protein 2; achaete-scute complex homolog 2; achaete-scute complex homolog 2 (Drosophila); achaete-scute complex homolog-like 2; achaete-scute complex-like 2; achaete-scute complex-like protein 2; achaete-scute family bHLH transcription factor 2; achaete-scute homolog 2; Ascl2; ASH2; ASH-2; bHLH transcription factor; Mash2; BHLHA45; Class A basic helix-loop-helix protein 45; H M bHLHa45; HASH2; long transient receptor potential-related channel 5; mammalian achaete scute homolog 2; mammalian achaete/scute homologue 2; MASH2; mASH-2 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 430 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q99929 | |
| ASCL2 | |
| Synthetic peptide directed towards the C-terminal of human ASCL2 (aa 121-170). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction