missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ASCIZ Antibody, PerCP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-79016PCP
This item is not returnable.
View return policy
Description
ASCIZ Polyclonal antibody specifically detects ASCIZ in Human samples. It is validated for Western Blot, Immunoprecipitation
Specifications
| ASCIZ | |
| Polyclonal | |
| Western Blot, Immunoprecipitation | |
| ASCIZ, ATM INteracting protein, ATM interactor, ATM/ATR-substrate CHK2-interacting zinc finger protein, ATM/ATR-Substrate Chk2-Interacting Zn++-finger protein, DKFZp779K1455, FLJ50270, FLJ61620, FLJ76795, KIAA0431, zinc finger protein 822, ZNF822 | |
| Maps to a region between residue 773 and 823 of human ATM Interactor using the numbering given in entry NP_056066.2 (GeneID 23300). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunoprecipitation | |
| PerCP | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 23300 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction