missing translation for 'onlineSavingsMsg'
Learn More

ASCIZ Antibody, Janelia Fluor™ 549, Novus Biologicals™

Product Code. 18400586 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18400586 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18400586 Supplier Novus Biologicals Supplier No. NBP281796JF549

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ASCIZ Polyclonal antibody specifically detects ASCIZ in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen ASCIZ
Applications Western Blot, ELISA, Immunofluorescence
Classification Polyclonal
Conjugate Janelia Fluor 549
Formulation 50mM Sodium Borate
Gene Alias ASCIZ, ATM INteracting protein, ATM interactor, ATM/ATR-substrate CHK2-interacting zinc finger protein, ATM/ATR-Substrate Chk2-Interacting Zn++-finger protein, DKFZp779K1455, FLJ50270, FLJ61620, FLJ76795, KIAA0431, zinc finger protein 822, ZNF822
Host Species Rabbit
Immunogen ASCIZ antibody was raised against a 19 amino acid peptide near the amino terminus of human ASCIZ. The immunogen is located within amino acids 200 - 250 of ASCIZ. Amino Acid Squence: HEIPAEHRDPPSKKRKMEN
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23300
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.