missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ASCIZ Antibody, Alexa Fluor™ 488, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-81796AF488
This item is not returnable.
View return policy
Description
ASCIZ Polyclonal antibody specifically detects ASCIZ in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunofluorescence
Specifications
| ASCIZ | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| 23300 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, ELISA, Immunofluorescence | |
| Alexa Fluor 488 | |
| ASCIZ, ATM INteracting protein, ATM interactor, ATM/ATR-substrate CHK2-interacting zinc finger protein, ATM/ATR-Substrate Chk2-Interacting Zn++-finger protein, DKFZp779K1455, FLJ50270, FLJ61620, FLJ76795, KIAA0431, zinc finger protein 822, ZNF822 | |
| ASCIZ antibody was raised against a 19 amino acid peptide near the amino terminus of human ASCIZ. The immunogen is located within amino acids 200 - 250 of ASCIZ. Amino Acid Squence: HEIPAEHRDPPSKKRKMEN | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction