missing translation for 'onlineSavingsMsg'
Learn More

ASCC3L1 Antibody, Novus Biologicals™

Product Code. 18394811 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18394811 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18394811 Supplier Novus Biologicals Supplier No. NBP287044

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ASCC3L1 Polyclonal specifically detects ASCC3L1 in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ASCC3L1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml
Formulation PBS, 2% Sucrose
Gene Alias ASCC3L1, BRR2, BRR2 homolog, FLJ11521, HELIC2, KIAA0788, RP33, small nuclear ribonucleoprotein 200kDa (U5), U5 small nuclear ribonucleoprotein 200 kDa helicase, U5 snRNP-specific 200 kDa protein, U5-200KD
Host Species Rabbit
Immunogen The immunogen is a synthetic peptide directed towards the N-terminal region of ASCC3L1
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23020
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.