missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ASCC3L1 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£151.00 - £359.00
Specifications
| Antigen | ASCC3L1 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18634962
|
Novus Biologicals
NBP2-92466-0.02ml |
0.02 mL |
£151.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18695572
|
Novus Biologicals
NBP2-92466-0.1ml |
0.1 mL |
£359.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ASCC3L1 Polyclonal antibody specifically detects ASCC3L1 in Human samples. It is validated for Western BlotSpecifications
| ASCC3L1 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| ASCC3L1, BRR2, BRR2 homolog, FLJ11521, HELIC2, KIAA0788, RP33, small nuclear ribonucleoprotein 200kDa (U5), U5 small nuclear ribonucleoprotein 200 kDa helicase, U5 snRNP-specific 200 kDa protein, U5-200KD | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1947-2136 of human SNRNP200 (NP_054733.2). QMVTQAMWSKDSYLKQLPHFTSEHIKRCTDKGVESVFDIMEMEDEERNALLQLTDSQIADVARFCNRYPNIELSYEVVDKDSIRSGGPVVVLVQLEREEEVTGPVIAPLFPQKREEGWWVVIGDAKSNSLISIKRLTLQQKAKVKLDFVAPATGAHNYTLYFMSDAYMGCDQEYKFSVDVKEAETDSDSD | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS with 50% glycerol, pH7.3. | |
| 23020 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title