missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
ASCC3L1 Polyclonal antibody specifically detects ASCC3L1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | ASCC3L1 |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Alexa Fluor 700 |
| Dilution | Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin |
| Formulation | 50mM Sodium Borate |
| Gene Alias | ASCC3L1, BRR2, BRR2 homolog, FLJ11521, HELIC2, KIAA0788, RP33, small nuclear ribonucleoprotein 200kDa (U5), U5 small nuclear ribonucleoprotein 200 kDa helicase, U5 snRNP-specific 200 kDa protein, U5-200KD |
| Host Species | Rabbit |
| Immunogen | The immunogen recognized by this antibody maps to a region between residue 400 and 450 of human Small Nuclear Ribonucleoprotein 200kDa using the numbering given in entry NP_054733.2 (GeneID 23020). |
| Purification Method | Immunogen affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?