missing translation for 'onlineSavingsMsg'
Learn More

ASCC3 Antibody - BSA Free, Novus Biologicals™

Product Code. 18689071 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.02 mL
0.1 mL
Unit Size:
0.01mL
0.02mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18689071 0.02 mL 0.02mL
18671992 0.1 mL 0.01mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18689071 Supplier Novus Biologicals Supplier No. NBP2926820.02ml

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ASCC3 Polyclonal antibody specifically detects ASCC3 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen ASCC3
Applications Western Blot, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500 - 1:1000, Immunocytochemistry/ Immunofluorescence 1:50-1:100
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias activating signal cointegrator 1 complex subunit 3, B630009I04Rik, HELIC1, helicase, ATP binding 1, p200, RNA helicase family
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-111 of human ASCC3 (NP_071374.1). MALPRLTGALRSFSNVTKQDNYNEEVADLKIKRSKLHEQVLDLGLTWKKIIKFLNEKLEKSKMQSINEDLKDILHAAKQIEVNCPFQKRRLDGKEEDEKMSRASDRFRGLR
Purification Method Affinity purified
Quantity 0.02 mL
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Gene ID (Entrez) 10973
Target Species Human, Mouse
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.