missing translation for 'onlineSavingsMsg'
Learn More

ASCC2 Antibody (OTI1E8), DyLight 594, Novus Biologicals™

Product Code. 18415537 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18415537 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18415537 Supplier Novus Biologicals Supplier No. NBP272273DL594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

ASCC2 Monoclonal antibody specifically detects ASCC2 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen ASCC2
Applications Flow Cytometry, Immunofluorescence
Classification Monoclonal
Clone OTI1E8
Conjugate DyLight 594
Formulation 50mM Sodium Borate
Gene Alias activating signal cointegrator 1 complex subunit 2, ASC 1 complex subunit P100, ASC-1 complex subunit p100, ASC1p100, DKFZp586O0223, FLJ21588, Trip4 complex subunit p100
Host Species Mouse
Immunogen Full length recombinant protein of human ASCC2 (NP_115580) produced in HEK293T cells.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84164
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.