missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ASCC1 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92631-0.02ml
This item is not returnable.
View return policy
Description
ASCC1 Polyclonal antibody specifically detects ASCC1 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| ASCC1 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| activating signal cointegrator 1 complex subunit 1, ASC-1 complex subunit p50, ASC1p50, CGI-18, Em:AC022392.3, RP11-150D20.4, Trip4 complex subunit p50 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-180 of human ASCC1 (NP_001185727.1). MEVLRPQLIRIDGRNYRKNPVQEQTYQHEEDEEDFYQGSMECADEPCDAYEVEQTPQGFRSTLRAPSLLYKHIVGKRGDTRKKIEMETKTSISIPKPGQDGEIVITGQHRNGVISARTRIDVLLDTFRRKQPFTHFLAFFLNEVEVQEGFLRFQEEVLAKCSMDHGVDSSIFQNPKKLHL | |
| 0.02 mL | |
| Signal Transduction | |
| 51008 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction