missing translation for 'onlineSavingsMsg'
Learn More

Ascarite(II), CO2 absorbent, 20-30 mesh, capacity: 40-50%, Thermo Scientific Chemicals

Ascarite(II), CO2 absorbent, 20-30 mesh, capacity: 40-50%, CAS Number-81133-20-2, 1310-73-2, 7631-86-9, 100g, Beige to Brown, Danger, GHS H Statement: Causes severe skin burns and eye damage.

Brand:  Thermo Scientific Acros 208090025

Code : NEW

Additional Details : CAS Number : 81133-20-2 Weight : 2.50000kg Transport : UN number : 1823 Chem class : 8 Pack group : II

 View more versions of this product

Product Code. 15224684

  • £1457.00 / 2.50kg
Estimated Shipment: 22-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Gas Adsorbent
Specifications

Specifications

Granular
Ascarite(II)™, CO2 absorbent, 20-30 mesh, capacity: 40-50%
81133-20-2,1310-73-2,7631-86-9
MFCD00132264
Particle size analysis: 20 to 30 mesh
Beige to Brown
20- to 30-mesh
2.5 kg
Glass Bottle
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Special Offers

Special Offers

RUO – Research Use Only