missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ASC2 Antibody (OTI9C10), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-72480CL1
This item is not returnable.
View return policy
Description
ASC2 Monoclonal antibody specifically detects ASC2 in Human samples. It is validated for Immunohistochemistry,Western Blot
Specifications
| ASC2 | |
| Monoclonal | |
| CoraFluor 1 | |
| ASC2PAAD-only protein 1, ASCI, POP1pyrin domain-containing protein 1, PYC1, PYD (pyrin domain) containing 1, pyrin domain containing 1, pyrin-domain containing protein 1, Pyrin-only protein 1 | |
| Full length human recombinant protein of human PYDC1 (NP_690865) produced in E.coli. | |
| 0.1 mL | |
| Immunology, Innate Immunity | |
| 260434 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 |
| Immunohistochemistry, Western Blot | |
| OTI9C10 | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction