Learn More
ASC1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA528305
Description
Recommended positive controls: Neuro2A, GL261, C8D30, NIH-3T3, BCL-1, Raw264.7. Predicted reactivity: Mouse (86%), Rat (85%), Rhesus Monkey (98%), Chimpanzee (100%), Bovine (87%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Transcription coactivator of nuclear receptors which functions in conjunction with CBP-p300 and SRC-1 and may play an important role in establishing distinct coactivator complexes under different cellular conditions. Plays a pivotal role in the transactivation of NF-kappa-B, SRF and AP1. Acts as a mediator of transrepression between nuclear receptor and either AP1 or NF-kappa-B. Plays a role in androgen receptor transactivation and in testicular function.
Specifications
| ASC1 | |
| Polyclonal | |
| Unconjugated | |
| Trip4 | |
| 4930558E03Rik; Activating signal cointegrator 1; Asc1; ASC-1; BB191711; HsT17391; SMABF1; thyroid hormone receptor interactor 4; thyroid receptor-interacting protein 4; transcription coactivator Asc1; TR-interacting protein 4; TRIP4; TRIP-4; ZC2HC5; zinc finger, C2HC5-type | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 56404, 9325 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
| Q15650, Q9QXN3 | |
| Trip4 | |
| Recombinant fragment corresponding to a region within amino acids 192 and 515 of Human ASC1 | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.