missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ASC1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£328.00 - £513.00
Specifications
| Antigen | ASC1 |
|---|---|
| Applications | Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18286401
|
Novus Biologicals
NBP2-58194 |
100 μL |
£513.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18644717
|
Novus Biologicals
NBP2-58194-25ul |
25 μL |
£328.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ASC1 Polyclonal specifically detects ASC1 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
| ASC1 | |
| Polyclonal | |
| Rabbit | |
| Apoptosis, Cancer, DNA Repair, Tumor Suppressors | |
| activating signal cointegrator 1, ASC-1, HsT17391, thyroid hormone receptor interactor 4, thyroid receptor interacting protein 4, Thyroid receptor-interacting protein 4, TR-interacting protein 4, TRIP-4 | |
| TRIP4 | |
| IgG | |
| Affinity Purified |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| Human | |
| 9325 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:KAFRSSGFGLEFNSFQHQLRIQDQEFQEGFDGGWCLSVHQPWASLLVRGIKRVEGRSWYTPHRGRLWIAATAKKPSPQEVSELQATYRLLRGKDVEFPNDYPSGCLLGCVDLIDCLSQKQFKEQFPDISQESDSPFVF | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title