Learn More
ASC/TMS1 Mouse anti-Human, Unconjugated, Clone: 2G12E6, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67494-1-IG-150UL
This item is not returnable.
View return policy
Description
The ASC/TMS1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human ASC/TMS1. This antibody recognizes human antigen. The ASC/TMS1 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| ASC/TMS1 | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human |
| Western Blot, Immunohistochemistry | |
| 2G12E6 | |
| Unconjugated | |
| 150 μL | |
| 29108 | |
| IgG1 |