missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ASC/TMS1 Antibody, DyLight 405, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-78978V
This item is not returnable.
View return policy
Description
ASC/TMS1 Polyclonal specifically detects ASC/TMS1 in Human, Mouse samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Western Blot (Negative).
Specifications
| ASC/TMS1 | |
| Polyclonal | |
| Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 1:40-1:100, Immunohistochemistry-Paraffin 1:200, Western Blot (Negative) | |
| Q9ULZ3 | |
| PYCARD | |
| This ASC/TMS1 Antibody was developed against a synthetic peptide made to a C-terminal portion of the human ASC/TMS1 protein (between residues 145-195) [Uniprot: Q9ULZ3] | |
| 0.1 mL | |
| Apoptosis, Cancer, Cell Biology, Inflammation, Innate Immunity | |
| 29108 | |
| Store at 4C in the dark. |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| DyLight 405 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| ASCMGC10332, CARD5TMS, caspase recruitment domain protein 5, Caspase recruitment domain-containing protein 5, hASC, PYD and CARD domain containing, PYD and CARD domain-containing protein, Target of methylation-induced silencing 1, TMS-1, TMS1apoptosis-associated speck-like protein containing a CARD | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction