missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ASC/TMS1 Antibody (1C3D3) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-61676
This item is not returnable.
View return policy
Description
ASC/TMS1 Monoclonal specifically detects ASC/TMS1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| ASC/TMS1 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunocytochemistry/Immunofluorescence 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000 | |
| ASCMGC10332, CARD5TMS, caspase recruitment domain protein 5, Caspase recruitment domain-containing protein 5, hASC, PYD and CARD domain containing, PYD and CARD domain-containing protein, Target of methylation-induced silencing 1, TMS-1, TMS1apoptosis-associated speck-like protein containing a CARD | |
| Mouse | |
| 21.6 kDa | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1C3D3 | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| PYCARD | |
| This ASC/TMS1 Antibody (1C3D3) was developed against purified recombinant fragment of human ASC/TMS1 (AA: 1-120) expressed in E. Coli. | |
| Protein G purified | |
| Apoptosis, Cancer, Cell Biology, Inflammation, Innate Immunity | |
| 29108 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction