missing translation for 'onlineSavingsMsg'
Learn More

ASB8 Antibody (OTI3A12), DyLight 594, Novus Biologicals™

Product Code. 18403137 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18403137 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18403137 Supplier Novus Biologicals Supplier No. NBP272400DL594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

ASB8 Monoclonal antibody specifically detects ASB8 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen ASB8
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI3A12
Conjugate DyLight 594
Formulation 50mM Sodium Borate
Gene Alias ankyrin repeat and SOCS box containing 8, ankyrin repeat and SOCS box protein 8, ankyrin repeat and SOCS box-containing 8, ASB-8, FLJ21255, MGC5540
Host Species Mouse
Immunogen Full length recombinant protein of human ASB8 (NP_077000) produced in E.coli.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 140461
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.