missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ASB6 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5107070
This item is not returnable.
View return policy
Description
Sequence homology: Cow: 93%; Dog: 93%; Guinea Pig: 93%; Horse: 93%; Human: 100%; Mouse: 100%; Rat: 100%; Zebrafish: 79%.
ASB6 belongs to a family of ankyrin repeat proteins that, along with four other protein families, contain a C-terminal SOCS box motif. Growing evidence suggests that the SOCS box, similar to the F-box, acts as a bridge between specific substrate-binding domains and the more generic proteins that comprise a large family of E3 ubiquitin protein ligases.The protein encoded by this gene belongs to a family of ankyrin repeat proteins that, along with four other protein families, contain a C-terminal SOCS box motif. Growing evidence suggests that the SOCS box, similar to the F-box, acts as a bridge between specific substrate-binding domains and the more generic proteins that comprise a large family of E3 ubiquitin protein ligases.
Specifications
| ASB6 | |
| Polyclonal | |
| Unconjugated | |
| ASB6 | |
| ankyrin repeat and SOCS box containing 6; ankyrin repeat and SOCS box protein 6; ankyrin repeat and SOCS box-containing 6; ASB6; ASB-6 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 140459 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide; pH 7.4 | |
| Q9NWX5 | |
| ASB6 | |
| Synthetic peptide directed towards the middle region of human ASB6 (aa 74-124). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction