missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ASB2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA529476
This item is not returnable.
View return policy
Description
Recommended positive controls: HL-60. Predicted reactivity: Mouse (93%), Rat (93%), Bovine (93%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The protein encoded by this gene is a member of the ankyrin repeat and SOCS box-containing (ASB) family of proteins. They contain ankyrin repeat sequence and SOCS box domain. The SOCS box serves to couple suppressor of cytokine signalling (SOCS) proteins and their binding partners with the elongin B and C complex, possibly targeting them for degradation. This gene is induced by all-trans retinoic acid. In myeloid leukemia cells, the expression of this encoded protein has been shown to induce growth inhibition and chromatin condensation. Multiple alternatively spliced transcript variants have been described for this gene but their full length sequences are not known.
Specifications
| ASB2 | |
| Polyclonal | |
| Unconjugated | |
| ASB2 | |
| 1110008E15Rik; ankyrin repeat and SOCS box containing 2; ankyrin repeat and SOCS box protein 2; ankyrin repeat and SOCS box-containing 2; ankyrin repeat and SOCS box-containing protein 2; ankyrin repeat and SOCS box-containing protein 2a; ankyrin repeat-containing SOCS box protein 2; Asb2; ASB-2; asb2 {ECO:0000312; EMBL:AAH85882.1}; MGC40044 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 51676, 65256 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
| Q8K0L0, Q96Q27 | |
| ASB2 | |
| Recombinant protein encompassing a sequence within the center region of human ASB2. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction