missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ASB2 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92391-0.1ml
This item is not returnable.
View return policy
Description
ASB2 Polyclonal antibody specifically detects ASB2 in Human, Rat samples. It is validated for Western Blot
Specifications
| ASB2 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| ankyrin repeat and SOCS box containing 2, ankyrin repeat and SOCS box protein 2, ankyrin repeat and SOCS box-containing protein 2a, ASB-2ankyrin repeat and SOCS box-containing 2, MGC40044 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 448-587 of human ASB2 (NP_057234.2). MDLGCDGEPCFSCLYGNGPHPPAPQPSSRFNDAPAADKEPSVVQFCEFVSAPEVSRWAGPIIDVLLDYVGNVQLCSRLKEHIDSFEDWAVIKEKAEPPRPLAHLCRLRVRKAIGKYRIKLLDTLPLPGRLIRYLKYENTQ | |
| 0.1 mL | |
| Signal Transduction | |
| 51676 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction