missing translation for 'onlineSavingsMsg'
Learn More

ASB17 Rabbit anti-Human, Rat, Polyclonal, Novus Biologicals™

Product Code. 18315734
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18315734 20 μg 20µL
18333032 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18315734 Supplier Novus Biologicals Supplier No. NBP31598720UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ASB17 Polyclonal antibody specifically detects ASB17 in Human, Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen ASB17
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS, 50% glycerol, pH7.3
Gene Alias ankyrin repeat and SOCS box containing 17, ankyrin repeat and SOCS box protein 17, ankyrin repeat and SOCS box-containing 17, Asb-17
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 78-177 of mouse ASB17 (NP_080034.2).
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 127247
Target Species Human, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.