missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ASB13 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA566419
This item is not returnable.
View return policy
Description
Immunogen sequence: KVKNVDLIEML IEFGGNIYAR DNRGKKPSDY TWSSSAPAKC FEYYEKTPLT LSQLCRVNLR KATGVRGLEK IAKLNI Highest antigen sequence identity to the following orthologs - mouse 96%, rat 96%.
The protein encoded by this gene is a member of the ankyrin repeat and SOCS box-containing family of proteins. They contain ankyrin repeat sequence and a SOCS box domain. The SOCS box serves to couple suppressor of cytokine signalling proteins and their binding partners with the elongin B and C complex, possibly targeting them for degradation. Multiple alternatively spliced transcript variants have been described for this gene, but their full-length sequences are not known.
Specifications
| ASB13 | |
| Polyclonal | |
| Unconjugated | |
| ASB13 | |
| 2210015B19Rik; 6430573K02Rik; ankyrin repeat and SOCS box containing 13; ankyrin repeat and SOCS box protein 13; ankyrin repeat and SOCS box-containing 13; ankyrin repeat and SOCS box-containing protein 13; ankyrin repeat domain-containing SOCS box protein 13; ankyrin repeat domain-containing SOCS box protein Asb-13; ASB13; Asb-13; C85285 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 79754 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8WXK3 | |
| ASB13 | |
| Recombinant Human ASB13. Recombinant protein control fragment (Product #RP-107091). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction