missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ASB13 Antibody (OTI7E10), Novus Biologicals Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-44956-30ul
This item is not returnable.
View return policy
Description
ASB13 Monoclonal antibody specifically detects ASB13 in Human,Mouse,Rat samples. It is validated for Western Blot
Specifications
| ASB13 | |
| Monoclonal | |
| Unconjugated | |
| PBS (pH 7.3) 1% BSA, 50% glycerol | |
| Mouse | |
| >95% | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI7E10 | |
| Western Blot 1:500 | |
| ankyrin repeat and SOCS box containing 13, ankyrin repeat and SOCS box protein 13, ankyrin repeat and SOCS box-containing 13, ankyrin repeat domain-containing SOCS box protein Asb-13, ASB-13, FLJ13134, MGC19879 | |
| Human recombinant protein fragment corresponding to amino acids 76-278 of human ASB13(NP_078977) produced in E.coli. | |
| 30 μL | |
| Signal Transduction | |
| 79754 | |
| Store at -20 degrees C. Avoid freeze/thaw cycles. | |
| Purified |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur