missing translation for 'onlineSavingsMsg'
Learn More

ASB13 Antibody (OTI4A1) [DyLight 594], Novus Biologicals™

Product Code. 30688049 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30688049 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30688049 Supplier Novus Biologicals Supplier No. NBP344955DL594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

ASB13 Monoclonal antibody specifically detects ASB13 in Human,Mouse,Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen ASB13
Applications Western Blot
Classification Monoclonal
Clone OTI4A1
Conjugate DyLight 594
Formulation 50mM Sodium Borate
Gene Alias ankyrin repeat and SOCS box containing 13, ankyrin repeat and SOCS box protein 13, ankyrin repeat and SOCS box-containing 13, ankyrin repeat domain-containing SOCS box protein Asb-13, ASB-13, FLJ13134, MGC19879
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 76-278 of human ASB13(NP_078977) produced in E.coli.
Purification Method >95%
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 79754
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.