missing translation for 'onlineSavingsMsg'
Learn More

ASB13 Antibody (OTI3F9) [DyLight 650], Novus Biologicals™

Product Code. 30689494 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30689494 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30689494 Supplier Novus Biologicals Supplier No. NBP344952C

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

ASB13 Monoclonal antibody specifically detects ASB13 in Human,Mouse,Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen ASB13
Applications Western Blot
Classification Monoclonal
Clone OTI3F9
Conjugate DyLight 650
Formulation 50mM Sodium Borate
Gene Alias ankyrin repeat and SOCS box containing 13, ankyrin repeat and SOCS box protein 13, ankyrin repeat and SOCS box-containing 13, ankyrin repeat domain-containing SOCS box protein Asb-13, ASB-13, FLJ13134, MGC19879
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 76-278 of human ASB13 (NP_078977) produced in E.coli.
Purification Method >95%
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 79754
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.