missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ASB1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA551946
This item is not returnable.
View return policy
Description
Immunogen sequence: LVDVKGQTAL YVAVVNGHLE STQILLEAGA DPNGSRHHRS TPVYHASRVG RADILKALIR YGADVDVNHH LTPDVQPRFS RRLTSLVVCP LYISAAYHNL QCFRLLLLAG ANPDFNCNGP VNTQGFYRGS PGCVMDAVL Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 95%.
The protein encoded by this gene contains an ankyrin repeat sequence and SOCS box domain. The SOCS box serves to couple suppressor of cytokine signaling (SOCS) proteins and their binding partners with the elongin B and C complex, targeting them for ubiquitination and degradation.
Specifications
| ASB1 | |
| Polyclonal | |
| Unconjugated | |
| ASB1 | |
| 1700029O08Rik; 1700054C17Rik; AI426616; AI851378; ankyrin repeat and SOCS box containing 1; ankyrin repeat and SOCS box protein 1; ankyrin repeat and SOCS box-containing 1; ankyrin repeat and SOCS box-containing protein 1; ankyrin repeat-containing SOCS box protein 1; ASB1; ASB-1; KIAA1146; LOW QUALITY PROTEIN: ankyrin repeat and SOCS box protein 1; mKIAA1146 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 51665 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9Y576 | |
| ASB1 | |
| Recombinant protein corresponding to Human ASB1. Recombinant protein control fragment (Product #RP-102064). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction