missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ ASB 14780
GREENER_CHOICE

Product Code. 18748016
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18748016 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18748016 Supplier Tocris Bioscience™ Supplier No. 6316/5

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent and selective cytosolic phospholipase A2 alpha (cPLA2α) inhibitor; active in vivo; orally bioavailable

ASB 14780 is a potent and selective cytosolic phospholipase A2 alpha (cPLA2α) inhibitor (IC50 = 20 nM). Selective for cPLA2α over secreted PLA2α (sPLA2α; exhibits no inhibition at 10 μM). Inhibits cPLA2α-dependent inflammatory responses in vitro in guinea pig and human whole-blood assays. Improves diet-induced liver injury and CCl4-induced hepatic fibrosis in vivo. Orally bioavailable.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-(3-Phenethyl-1-(4-phenoxyphenyl)-1 H-indol-5-yl)propanoic acid tris salt
CAS 1069046-00-9
Quantity 5 mg
Target Phospholipase Inhibitors
Molecular Formula C31H27NO3.C4H11NO3
Purity 0.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.