missing translation for 'onlineSavingsMsg'
Learn More

Asaraldehyde, MedChemExpress

Product Code. 16454386
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16454386 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16454386 Supplier Medchem Express Supplier No. HY100580

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress Asarylaldehyde (Asaronaldehyde), a COX-2 inhibitor, significantly inhibits cyclooxygenase II (COX-2) activity with an IC50 value of 100 μg/mL.

Asarylaldehyde (Asaronaldehyde), a COX-2 inhibitor, significantly inhibits cyclooxygenase II (COX-2) activity with an IC50 value of 100 μg/mL.

Specifications

Chemical Name or Material Asaraldehyde
CAS 4460-86-0
Color White to Off-White
Purity Grade Notes Research
Molecular Formula C10H12O4
Quantity 100 mg
Synonym Asaronaldehyde Asaraldehyde 2,4,5-trimethoxy-Benzaldehyde
Solubility Information DMSO : 100 mg/mL (509.68 mM; Need ultrasonic) ∣Ethanol : 50 mg/mL (254.84 mM; Need ultrasonic) ∣H2O : 0.14 mg/mL (0.71 mM; Need ultrasonic)
SMILES O=CC1=CC(OC)=C(OC)C=C1OC
Molecular Weight (g/mol) 196.2
Formula Weight 196.2
Percent Purity 97.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.