missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ASAP3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5110863
This item is not returnable.
View return policy
Description
Immunogen sequence: EAQLPSHGGP KPSAESDMGT RRDYIMAKYV EHRFARRCTP EPQRLWTAIC NRDLLSVLEA FA Highest antigen sequence indentity to the following orthologs: Mouse - 79%, Rat - 82%.
This gene encodes a member of a subfamily of ADP-ribosylation factor(Arf) GTPase-activating proteins that contain additional ankyrin repeat and pleckstrin homology domains. The Arf GAP domain of this protein catalyzes the hydrolysis of GTP bound to Arf proteins. The encoded protein promotes cell differentiation and migration and has been implicated in cancer cell invasion. Alternative splicing results in multiple transcript variants.
Specifications
| ASAP3 | |
| Polyclonal | |
| Unconjugated | |
| Asap3 | |
| 9430088F20Rik; ACAP4; ARF6 GTPase-activating protein; Arf-GAP with SH3 domain, ANK repeat and PH domain-containing protein 3; ArfGAP with SH3 domain, ankyrin repeat and PH domain 3; ArfGAP with SH3 domain, ankyrin repeat and PH domain 3 1; Asap3; centaurin, beta 6; CENTB6; DDEFL1; development and differentiation enhancing factor-like 1; development and differentiation-enhancing factor-like 1; Gm140; Protein up-regulated in liver cancer 1; UPLC1; up-regulated in liver cancer 1 (UPLC1) | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 55616 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8TDY4 | |
| Asap3 | |
| Recombinant Protein corresonding to Human ASAP3. Recombinant protein control fragment (Product #RP-91722). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction