missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ASAP1 Polyclonal Antibody

Brand: Thermo Scientific PA5114882
This item is not returnable.
View return policy
Description
Specifications
| ASAP1 | |
| Polyclonal | |
| Unconjugated | |
| ASAP1 | |
| 130 kDa phosphatidylinositol 4,5-biphosphate-dependent ARF1 GTPase-activating protein; 130 kDa phosphatidylinositol 4,5-bisphosphate-dependent ARF1 GTPase-activating protein; ADP-ribosylation factor-directed GTPase-activating protein 1; AMAP1; ARF GTPase-activating protein 1; arf-GAP with SH3 domain, ANK repeat and PH domain-containing protein 1; ArfGAP with SH3 domain, ankyrin repeat and PH domain 1; ArfGAP with SH3 domain, ankyrin repeat and PH domain1; Asap1; AV239055; centaurin, beta 4; CENTB4; Ddef1; DEF-1; development and differentiation enhancing factor 1; development and differentiation-enhancing factor 1; differentiation enhancing factor 1; differentiation-enhancing factor 1; Kiaa1249; mKIAA1249; PAG2; PAP; PIP2-dependent ARF1 GAP; s19; Shag1; src SH3 binding protein; ZG14P | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 13196, 314961, 50807 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide | |
| Q1AAU6, Q9QWY8, Q9ULH1 | |
| ASAP1 | |
| A synthesized peptide derived from human ASAP1(Accession Q9ULH1), corresponding to amino acid residues K508-S558. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction