missing translation for 'onlineSavingsMsg'
Learn More

ASAM Antibody [Janelia Fluor™ 525], Novus Biologicals™

Product Code. 30731213 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30731213 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30731213 Supplier Novus Biologicals Supplier No. AF7356JF525

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

ASAM Polyclonal antibody specifically detects ASAM in Human,Mouse samples. It is validated for Flow Cytometry,Western Blot,CyTOF
TRUSTED_SUSTAINABILITY

Specifications

Antigen ASAM
Applications Flow Cytometry, Western Blot, CyTOF
Classification Polyclonal
Conjugate Janelia Fluor 525
Formulation 50mM Sodium Borate
Gene Alias ACAMCoxsackie- and adenovirus receptor-like membrane protein, Adipocyte adhesion molecule, adipocyte-specific adhesion molecule, ASAMCAR-like membrane protein, CXADR-like membrane protein, FLJ22415
Host Species Sheep
Immunogen Chinese hamster ovary cell line CHO-derived recombinant mouse ASAM, Thr18-Ala234, Accession # Q8R373
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 79827
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.