missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ASAM Antibody (030), DyLight 650, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90635C
This item is not returnable.
View return policy
Description
ASAM Monoclonal specifically detects ASAM in Mouse samples. It is validated for ELISA.
Specifications
| ASAM | |
| Monoclonal | |
| DyLight 650 | |
| ACAMCoxsackie- and adenovirus receptor-like membrane protein, Adipocyte adhesion molecule, adipocyte-specific adhesion molecule, ASAMCAR-like membrane protein, CXADR-like membrane protein, FLJ22415 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Mouse ASAM (Q8R373-1; Met 1-Met 232). | |
| 0.1 mL | |
| Signal Transduction | |
| 79827 | |
| Mouse | |
| Purified |
| ELISA | |
| 30 | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Has cross-reactivity in ELISA with: Human ASAM | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction