missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ASAHL/N-acylethanolamine-hydrolyzing Acid Amidase Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£381.00
Specifications
| Antigen | ASAHL/N-acylethanolamine-hydrolyzing Acid Amidase |
|---|---|
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Regulatory Status | RUO |
Description
ASAHL/N-acylethanolamine-hydrolyzing Acid Amidase Polyclonal specifically detects ASAHL/N-acylethanolamine-hydrolyzing Acid Amidase in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| ASAHL/N-acylethanolamine-hydrolyzing Acid Amidase | |
| Unconjugated | |
| RUO | |
| Acid ceramidase-like protein, ASAHL, ASAH-like protein, EC 3.5.1.-, N-acylethanolamine acid amidase, N-acylethanolamine-hydrolyzing acid amidase, N-acylsphingosine amidohydrolase (acid ceramidase)-like, N-acylsphingosine amidohydrolase-like, PLT | |
| NAAA | |
| IgG | |
| 37 kDa |
| Polyclonal | |
| Rabbit | |
| Q02083 | |
| 27163 | |
| Synthetic peptides corresponding to ASAHL The peptide sequence was selected from the middle region of ASAHL. Peptide sequence VSWLIRATLSESENFEAAVGKLAKTPLIADVYYIVGGTSPREGVVITRNR. | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title