missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ASAH2 Polyclonal Antibody

Sheep Polyclonal Antibody
Brand: Invitrogen™ PA548033
This item is not returnable.
View return policy
Description
In direct ELISAs, approximately 15% cross-reactivity with recombinant human ASAH2 is observed. Reconstitute at 0.2 mg/mL in sterile PBS.
Sphingolipids are hydrolyzed by ceramidases to yield sphingosine and fatty acids. These ceramidases are classified according to the pH range that supports their optimal activity. ASAH2 is a neutral ceramidase and key regulator of sphingolipid signaling metabolites at the cell surface, catalyzing the hydrolysis of the N-acyl linkage of ceramide at an optimal pH of 6.5-8.5. ASAH2 is a type II integral membrane protein that can be cleaved to yield a soluble secreted protein and acts as a repressor of apoptosis both by reducing C16-ceramide, thereby preventing ceramide-induced apoptosis, and generating sphingosine. Sphingosine exerts both mitogenic and apoptosis-inducing activities, and its phosphorylated form functions as an intra- and intercellular second messenger. ASAH2 is ubiquitously expressed primarily expressed with higher levels in the intestine, kidney, skeletal muscle and heart. Recent studies indicate that ASAH2 encoded neutral ceramidase is a key enzyme for the catabolism of dietary sphingolipids and regulates the levels of bioactive sphingolipid metabolites in the intestinal tract.
Specifications
| ASAH2 | |
| Polyclonal | |
| Unconjugated | |
| Asah2 | |
| acylsphingosine deacylase 2; AI585898; Asah2; BCDase; hCD; HNAC1; LCDase; mitochondrial ceramidase; N-acylsphingosine amidohydrolase (non-lysosomal ceramidase) 2; N-acylsphingosine amidohydrolase 2; NCDase; N-CDase; Neutral ceramidase; Neutral ceramidase soluble form; neutral/alkaline ceramidase; non-lysosomal ceramidase | |
| Sheep | |
| Antigen Affinity Chromatography | |
| RUO | |
| 54447 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Immunoprecipitation, Western Blot | |
| 0.2 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| Q9JHE3 | |
| Asah2 | |
| Mouse myeloma cell line NS0-derived recombinant mouse ASAH2/N-acylsphingosine Amidohydrolase-2 Thr34-Thr756. | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction