missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ASAH2/N-acylsphingosine Amidohydrolase-2 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76360
This item is not returnable.
View return policy
Description
ASAH2/N-acylsphingosine Amidohydrolase-2 Polyclonal specifically detects ASAH2/N-acylsphingosine Amidohydrolase-2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA.
Specifications
| ASAH2/N-acylsphingosine Amidohydrolase-2 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| ASAH2 N-acylsphingosine amidohydrolase (non-lysosomal ceramidase) 2, BCDase, HNAC1, LCDase, NCDase, N-CDase | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 56624 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, ELISA | |
| 1 mg/mL | |
| Western Blot 1-2 μg/mL, ELISA | |
| NP_063946 | |
| ASAH2 | |
| Antibody was raised against a 17 amino acid synthetic peptide near the carboxy terminus of the human ASAH2. The immunogen is located within the last 50 amino acids of ASAH2. Amino Acid Squence: YRIRYFGHNRKQDILK | |
| 0.1 mg | |
| Primary | |
| Human, Mouse, Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction