missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ASAH2/N-acylsphingosine Amidohydrolase-2 Antibody (2), Alexa Fluor™ 405, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90347AF405
This item is not returnable.
View return policy
Description
ASAH2/N-acylsphingosine Amidohydrolase-2 Monoclonal specifically detects ASAH2/N-acylsphingosine Amidohydrolase-2 in Mouse samples. It is validated for ELISA.
Specifications
| ASAH2/N-acylsphingosine Amidohydrolase-2 | |
| Monoclonal | |
| Alexa Fluor 405 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 56624 | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA | |
| 2 | |
| ELISA | |
| ASAH2 N-acylsphingosine amidohydrolase (non-lysosomal ceramidase) 2, BCDase, HNAC1, LCDase, NCDase, N-CDase | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Mouse ASAH2/N-acylsphingosine Amidohydrolase-2 (Thr 34-Thr 756; NP_061300.1). | |
| 0.1 mL | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction