missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ ASAH1 HEK293 Cell Transient Overexpression Lysate (Nondenatured)
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
Specifications
Specifications
| Cell Line | HEK293 |
| For Use With (Application) | Immunoprecipitation, Western Blot |
| Description | ASAH1 HEK293 Cell Transient Overexpression Lysate(Non-Denatured) |
| Gene Accession No. | NM_177924 |
| Lysis Buffer | Modified RIPA Lysis Buffer:50 mM Tris-HCl pH 7.4, 150 mM NaCl, 1mM EDTA, 1% Triton X-100, 0.1% SDS, 1% Sodium deoxycholate, 1mM PMSF. ;Modified RIPA Lysis Buffer:50 mM Tris-HCl pH 7.4, 150 mM NaCl, 1mM EDTA, 1% Triton X-100, 0.1% SDS, 1% Sodium deoxycholate, 1mM PMSF. |
| Molecular Weight (g/mol) | Theoretical M.W. 45kDa |
| Plasmid | pCMV-ASAH1 full length |
| Quality Control Testing | Transient overexpression cell lysate was tested with anti-ASAH1 antibody (Mfr. No. H00000427-M01 ) by Western Blot. |
| Quantity | 100 μg |
| Storage Buffer | In modified RIPA Lysis Buffer. |
| Show More |
Safety and Handling
Product Identifier
- 100UG ASAH1 HEK293 Cell Transient Overexpression L ysate(Non-Denatured)
- Warning
- Aspiration toxicity Category 1
- Flammable liquid Category 3
- H226-Flammable liquid and vapour.
- H304-May be fatal if swallowed and enters airways.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- MIXTURE LIST-Contains : Dodecane
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction