missing translation for 'onlineSavingsMsg'
Learn More

ASAH1 Antibody (1A7), Novus Biologicals™

Product Code. 18318847 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18318847 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18318847 Supplier Novus Biologicals Supplier No. H00000427M02

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

ASAH1 Monoclonal antibody specifically detects ASAH1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen ASAH1
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 1A7
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_808592
Gene Alias ACDase, ACEC 3.5.1.23, Acid CDase, acid ceramidase, Acylsphingosine deacylase, ASAH, FLJ21558, FLJ22079, N-acylsphingosine amidohydrolase, N-acylsphingosine amidohydrolase (acid ceramidase) 1, PHP, PHP32N-acylsphingosine amidohydrolase (acid ceramidase), Putative 32 kDa heart protein
Host Species Mouse
Immunogen ASAH1 (NP_808592, 25 a.a. ~ 124 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. PPWTEDCRKSTYPPSGPTYRGAVPWYTINLDLPPYKRWHELMLDKAPMLKVIVNSLKNMINTFVPSGKVMQVVDEKLPGLLGNFPGPFEEEMKGIAAVTD
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 427
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG3 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.