missing translation for 'onlineSavingsMsg'
Learn More

AS8351, MedChemExpress

Product Code. 16434446
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16434446

Brand: Medchem Express HY100744

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress AS8351 (NSC51355) is a KDM5B inhibitor, which can induce and sustain active chromatin marks to facilitate the induction of cardiomyocyte-like cells.

AS8351 (NSC51355) is a KDM5B inhibitor, which can induce and sustain active chromatin marks to facilitate the induction of cardiomyocyte-like cells.

Specifications

Chemical Name or Material AS8351
CAS 796-42-9
Color Yellow
Purity Grade Notes Research
Molecular Formula C17H13N3O2
Quantity 100 mg
Synonym NSC51355
Solubility Information DMSO : 150 mg/mL (514.93 mM; Need ultrasonic and warming)
SMILES O=C(C1=CC=NC=C1)N/N=C/C2=C3C=CC=CC3=CC=C2O
Molecular Weight (g/mol) 291.3
Formula Weight 291.3
Percent Purity 99.89%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.